1-Decanethiol 96%

Supplier: Ambeed
Danger

Synonyms: n-Decanethiol, 1-Mercaptodecane, n-Decyl mercaptan

A444327-25G A444327-5G
77240-994EA 10 USD
77240-994 77240-996
1-Decanethiol 96%
1-Decanethiol

Caution: for Research Use Only

Formula: C₁₀H₂₂S
MDL Number: MFCD00004884
CAS Number: 143-10-2
UN: 3334
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR