GANT58 98%

Supplier: Ambeed
Warning

Synonyms: 4,4',4'',4'''-(Thiophene-2,3,4,5-tetrayl)tetrapyridine, 4-(2,4,5-Tri(pyridin-4-yl)thiophen-3-yl)pyridine, NSC 75503, Hh/Gli Antagonist, GANT 58

A430364-100MG A430364-1G A430364-250MG A430364-50MG
77252-452EA 37 USD
77252-452 77252-454 77252-456 77252-458
GANT58 98%
GANT58

Caution: for Research Use Only

Formula: C₂₄H₁₆N₄S
MW: 392.4756 g/mol
MDL Number: MFCD01103196
CAS Number: 64048-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR