Dichlorobis[2-(di-i-propylphosphino)ethylamine]ruthenium(II) 98%

Supplier: Ambeed
Warning

A422969-100MG A422969-1G A422969-250MG
77252-338EA 34 USD
77252-338 77252-340 77252-342
Dichlorobis[2-(di-i-propylphosphino)ethylamine]ruthenium(II) 98%
Dichlorobis[2-(di-i-propylphosphino)ethylamine]ruthenium(II)

Caution: for Research Use Only

Formula: C₁₆H₄₀Cl₂N₂P₂Ru
MDL Number: MFCD07782005
CAS Number: 1092372-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR