2'-Bromo-4'-fluoroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromo-4-fluorophenyl)ethanone

A387659-100G A387659-10G A387659-1G A387659-25G A387659-5G
77240-466EA 191 USD
77240-466 77240-468 77240-470 77240-472 77240-474
2'-Bromo-4'-fluoroacetophenone 98%
2'-Bromo-4'-fluoroacetophenone

Caution: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD00077464
CAS Number: 1006-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR