2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-Methylbut-3-yn-2-yloxy)oxane
A382248-100G
A382248-1G
A382248-250MG
A382248-5G
77278-538EA
639
USD
77278-538
77278-540
77278-542
77278-544
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran 97%
2-((2-Methylbut-3-yn-2-yl)oxy)tetrahydro-2H-pyran
Caution: for Research Use Only
Formula:
C₁₀H₁₆O₂ MW: 168.23 g/mol |
MDL Number:
MFCD00090526 CAS Number: 27943-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...