2-Adamantanol 98%

Supplier: Ambeed
Warning

A369281-25G A369281-5G A369281-100G
77251-742EA 33 USD
77251-742 77251-744 77746-742
2-Adamantanol 98%
2-Adamantanol

Caution: for Research Use Only

Formula: C₁₀H₁₆O
MDL Number: MFCD00074744
CAS Number: 700-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR