α,α,α-Trifluoro-p-toluic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Trifluoromethyl)benzoic acid

A329837-1G A329837-25G A329837-500G A329837-5G A329837-100G A329837-1000G
77239-750EA 5 USD
77239-750 77239-752 77239-754 77239-756 77550-926 77746-645
α,α,α-Trifluoro-p-toluic acid 97%
α,α,α-Trifluoro-p-toluic acid

Caution: for Research Use Only

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
MDL Number: MFCD00002562
CAS Number: 455-24-3
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR