2',3'-Dideoxycytidine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-1-((2R,5S)-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Zalcitabine

A310205-10MG A310205-1MG A310205-50MG A310205-1G A310205-100MG A310205-5G
77250-946EA 12.36 USD
77250-946 77250-948 77250-950 77601-402 77616-352 77746-593
2',3'-Dideoxycytidine 98%
2',3'-Dideoxycytidine

Caution: for Research Use Only

Formula: C₉H₁₃N₃O₃
MW: 211.22 g/mol
MDL Number: MFCD00012188
CAS Number: 7481-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR