N4-Acetylcytosine 98%

Supplier: Ambeed
Warning

A287573-1000G A287573-100G A287573-10G A287573-25G A287573-500G A287573-5G
77277-104EA 135 USD
77277-104 77277-106 77277-108 77277-110 77277-112 77277-114
N4-Acetylcytosine 98%
N4-Acetylcytosine

Caution: for Research Use Only

Formula: C₆H₇N₃O₂
MDL Number: MFCD00134466
CAS Number: 14631-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR