4-Iodo-3-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A284217-10G A284217-1G A284217-250MG A284217-25G A284217-5G
77289-698EA 25 USD
77289-698 77289-700 77289-702 77289-704 77289-706
4-Iodo-3-nitrobenzoic acid 97%
4-Iodo-3-nitrobenzoic acid

Caution: for Research Use Only

Formula: C₇H₄INO₄
MDL Number: MFCD00502399
CAS Number: 35674-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR