2-Phenylquinoline 97%

Supplier: Ambeed
Warning

A283805-10G A283805-1G A283805-5G
77250-546EA 55.99 USD
77250-546 77250-548 77250-550
2-Phenylquinoline 97%
2-Phenylquinoline

Caution: for Research Use Only

Formula: C₁₅H₁₁N
MDL Number: MFCD00011568
CAS Number: 612-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR