1-Nonanesulfonic acid sodium salt 99%

Supplier: Ambeed
Warning

Synonyms: Sodium 1-nonanesulfonate, Sodium nonane-1-sulfonate, Sodium 1-nonanesulphonate, Sodium nonane-1-sulphonate

A263102-10G A263102-1G A263102-25G
77238-542EA 27.81 USD
77238-542 77238-544 77238-546
1-Nonanesulfonic acid sodium salt 99%
1-Nonanesulphonic acid sodium salt

Caution: for Research Use Only

Formula: C₉H₁₉NaO₃S
MW: 230.30 g/mol
MDL Number: MFCD00025010
CAS Number: 35192-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR