4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-3,5-difluorophenyl 4-Amylbenzoate, 4-Cyano-3,5-difluorophenyl 4-Pentylbenzoate, 4-Amylbenzoic acid 4-Cyano-3,5-difluorophenyl ester, 4-Pentylbenzoic acid 4-Cyano-3,5-difluorophenyl ester

A235242-1G A235242-250MG A235242-5G A235242-25G
77288-544EA 13 USD
77288-544 77288-546 77288-548 77748-406
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate 97%
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate

Caution: for Research Use Only

Formula: C₁₉H₁₇F₂NO₂
MDL Number: MFCD11053375
CAS Number: 123843-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR