Dimethyl-3-cyclopentene-1,1-dicarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Dimethyl cyclopent-3-ene-1,1-dicarboxylate

A228430-100G A228430-1G A228430-25G A228430-5G
77275-702EA 362 USD
77275-702 77275-704 77275-706 77275-708
Dimethyl-3-cyclopentene-1,1-dicarboxylate 98%
Dimethyl-3-cyclopentene-1,1-dicarboxylate

Caution: for Research Use Only

Formula: C₉H₁₂O₄
MW: 184.19 g/mol
MDL Number: MFCD00154460
CAS Number: 84646-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR