2,3,4-Trifluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,3,4-Trifluorobenzeneboronic acid, Dihydroxy(2,3,4-trifluorophenyl)borane, (2,3,4-Trifluorophenyl)metaboric acid

A222388-10G A222388-25G A222388-1G
77288-244EA 23 USD
77288-244 77288-246 77748-360
2,3,4-Trifluorophenylboronic acid 98%
2,3,4-Trifluorophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₄BF₃O₂
MDL Number: MFCD01863168
CAS Number: 226396-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR