4-Bromomethyl-2-cyanobiphenyl 95%

Supplier: Ambeed
Danger

Synonyms: 4'-(Bromomethyl)-[1,1'-biphenyl]-2-carbonitrile

A215394-1000G A215394-100G A215394-10G A215394-500G A215394-5G
77275-292EA 165 USD
77275-292 77275-294 77275-296 77275-298 77275-300
4-Bromomethyl-2-cyanobiphenyl 95%
4-Bromomethyl-2-cyanobiphenyl

Caution: for Research Use Only

Formula: C₁₄H₁₀BrN
MW: 272.14 g/mol
MDL Number: MFCD00671503
CAS Number: 114772-54-2
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR