4'-Bromo-2,2,2-trifluoroacetophenone 98%

Supplier: Ambeed
Warning

A207792-10G A207792-1G A207792-25G A207792-500G A207792-5G A207792-100G
77287-848EA 34 USD
77287-848 77287-850 77287-852 77287-854 77287-856 77748-303
4'-Bromo-2,2,2-trifluoroacetophenone 98%
4'-Bromo-2,2,2-trifluoroacetophenone

Caution: for Research Use Only

Formula: C₈H₄BrF₃O
MDL Number: MFCD00191862
CAS Number: 16184-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR