4-Acetylbenzenesulfonamide 95%
Supplier: Ambeed
|
Warning
|
A197648-100MG
A197648-1G
A197648-250MG
A197648-25G
A197648-5G
77287-596EA
11.33
USD
77287-596
77287-598
77287-600
77287-602
77748-253
4-Acetylbenzenesulfonamide 95%
4-Acetylbenzenesulfonamide
Caution: for Research Use Only
Formula:
C₈H₉NO₃S |
MDL Number:
MFCD00792524 CAS Number: 1565-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...