4-Carboxy-3-chlorophenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Carboxy-3-chlorobenzeneboronic acid
A196413-10G
A196413-1G
A196413-250MG
A196413-25G
77274-626EA
37
USD
77274-626
77274-628
77274-630
77274-632
4-Carboxy-3-chlorophenylboronic acid 98%
4-Carboxy-3-chlorophenylboronic acid
Caution: for Research Use Only
Formula:
C₇H₆BClO₄ MW: 200.39 g/mol |
MDL Number:
MFCD06656268 CAS Number: 136496-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...