4-(2-Hydroxyethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A179583-10G A179583-1G A179583-250MG A179583-25G A179583-5G
77287-134EA 78 USD
77287-134 77287-136 77287-138 77287-140 77287-142
4-(2-Hydroxyethyl)benzonitrile 98%
4-(2-Hydroxyethyl)benzonitrile

Caution: for Research Use Only

Formula: C₉H₉NO
MDL Number: MFCD00052461
CAS Number: 69395-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR