2-Bromo-6-iodopyridine 97%

Supplier: Ambeed
Warning

A179344-250MG A179344-5G A179344-1G
77260-168EA 10 USD
77260-168 77260-170 77746-167
2-Bromo-6-iodopyridine 97%
2-Bromo-6-iodopyridine

Caution: for Research Use Only

Formula: C₅H₃BrIN
MW: 283.89
MDL Number: MFCD08059557
CAS Number: 234111-08-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR