6-Chlorooxindole 98%

Supplier: Ambeed
Warning

A171831-100G A171831-10G A171831-25G A171831-5G
77273-736EA 67 USD
77273-736 77273-738 77273-740 77273-742
6-Chlorooxindole 98%
6-Chlorooxindole

Caution: for Research Use Only

Formula: C₈H₆ClNO
MDL Number: MFCD00209962
CAS Number: 56341-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR