2,3,4,5-Tetrafluorobenzamide 95%

Supplier: Ambeed
Warning

A157787-100G A157787-25G
77247-548EA 390.01 USD
77247-548 77247-550
2,3,4,5-Tetrafluorobenzamide 95%
2,3,4,5-Tetrafluorobenzamide

Caution: for Research Use Only

Formula: C₇H₃F₄NO
MDL Number: MFCD01631457
CAS Number: 16582-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR