2-Bromoisobutyramide 98%

Supplier: Ambeed
Danger

A146640-10G A146640-1G A146640-25G A146640-5G A146640-100G
77259-044EA 8 USD
77259-044 77259-046 77259-048 77259-050 77746-031
2-Bromoisobutyramide 98%
2-Bromoisobutyramide

Caution: for Research Use Only

Formula: C₄H₈BrNO
MDL Number: MFCD08234837
CAS Number: 7462-74-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR