R788 disodium salt 98+%

Supplier: Ambeed
Warning

Synonyms: Fostamatinib Disodium, Sodium (6-((5-fluoro-2-((3,4,5-trimethoxyphenyl)amino)pyrimidin-4-yl)amino)-2,2-dimethyl-3-oxo-2,3-dihydro-4H-pyrido[3,2-b][1,4]oxazin-4-yl)methyl phosphate, Fostamatinib

A144826-10MG A144826-1MG A144826-50MG A144826-5MG
77235-142EA 37 USD
77235-142 77235-144 77235-146 77235-148
R788 disodium salt 98+%
R788 disodium salt

Caution: for Research Use Only

Formula: C₂₃H₂₄FN₆O₉P·2Na
MDL Number: MFCD15146370
CAS Number: 1025687-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR