Aloe emodin 97%

Supplier: Ambeed
Warning

Synonyms: Aloe-emodin, 3-Hydroxymethylchrysazine, 1,8-Dihydroxy-3-(hydroxymethyl)anthracene-9,10-dione, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone

A142364-250MG A142364-25MG
77235-034EA 10 USD
77235-034 77235-036
Aloe emodin 97%
Aloe emodin

Caution: for Research Use Only

Formula: C₁₅H₁₀O₅
MW: 270.24 g/mol
MDL Number: MFCD00017373
CAS Number: 481-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR