7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Methoxy-8-(3-methyl-2-butenyl)coumarin, 7-Methoxy-8-(3-methylbut-2-en-1-yl)-2H-chromen-2-one, Osthole
A141553-10G
A141553-1G
A141553-1MG
A141553-250MG
A141553-25G
A141553-5G
A141553-100G
77258-820EA
73
USD
77258-820
77258-822
77258-824
77258-826
77258-828
77258-830
77747-996
7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one 98%
7-Methoxy-8-(3-methyl-2-buten-1-yl)-2H-chromen-2-one
Caution: for Research Use Only
Formula:
C₁₅H₁₆O₃ MW: 244.2857 g/mol |
MDL Number:
MFCD00076049 CAS Number: 484-12-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...