1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 98%

Supplier: Ambeed
Warning

Synonyms: 1-O-Acetyl-2,3,5-Tri-O-benzoyl-beta-D-Ribofuranose, (2S,3R,4R,5R)-2-Acetoxy-5-((benzoyloxy)methyl)tetrahydrofuran-3,4-diyl dibenzoate, β-D-Ribofuranose 1-Acetate 2,3,5-Tribenzoate, 2,3,5-Tri-o-Benzoyl-1-o-Acetyl-β-D-Ribofuranose, 1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate

A139103-100G A139103-10G A139103-500G A139103-25G A139103-5G
77285-658EA 42 USD
77285-658 77285-660 77285-662 77315-774 77615-106
1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose 98%
1-O-Acetyl-2,3,5-tri-O-benzoyl-beta-D-ribofuranose

Caution: for Research Use Only

Formula: C₂₈H₂₄O₉
MW: 504.5 g/mol
MDL Number: MFCD00005357
CAS Number: 6974-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR