3-(3-Trifluoromethylphenyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(3-(Trifluoromethyl)phenyl)propanoic acid, 3-[3-(Trifluoromethyl)phenyl]propionic acid, 3-(Trifluoromethyl)hydrocinnamic acid

A134956-100G A134956-10G A134956-1G A134956-25G A134956-5G
77272-172EA 206 USD
77272-172 77272-174 77272-176 77272-178 77272-180
3-(3-Trifluoromethylphenyl)propionic acid 98%
3-(3-Trifluoromethylphenyl)propionic acid

Caution: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
MDL Number: MFCD00090487
CAS Number: 585-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR