2-Hydroxyisonicotinic acid 97%

Supplier: Ambeed
Warning

A131542-1G A131542-5G
77272-026EA 9 USD
77272-026 77272-028
2-Hydroxyisonicotinic acid 97%
2-Hydroxyisonicotinic acid

Caution: for Research Use Only

Formula: C₆H₅NO₃
MDL Number: MFCD01318566
CAS Number: 22282-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR