L(+)-Ornithine hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: H-Orn-OH Hydrochloride, H-Orn-OH·HCl

A128208-10G A128208-250MG A128208-25G A128208-500G A128208-1000G A128208-100G
77271-868EA 6 USD
77271-868 77271-870 77271-872 77271-874 77615-002 77615-004
L(+)-Ornithine hydrochloride 95%
L(+)-Ornithine hydrochloride

Caution: for Research Use Only

Formula: C₅H₁₂N₂O₂·HCl
MW: 168.62 g/mol
MDL Number: MFCD00064562
CAS Number: 3184-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR