6-Chloropyridine-2,3-diamine 98%

Supplier: Ambeed
Warning

A127746-100G A127746-1G A127746-5G A127746-10G
77246-394EA 467 USD
77246-394 77246-396 77246-398 77566-482
6-Chloropyridine-2,3-diamine 98%
6-Chloropyridine-2,3-diamine

Caution: For research use only.

Formula: C₅H₆ClN₃
MDL Number: MFCD00209966
CAS Number: 40851-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR