6-Chloropurine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine
A127343-100G
A127343-10G
A127343-25G
A127343-500G
A127343-5G
A127343-1000G
77271-840EA
30
USD
77271-840
77271-842
77271-844
77271-846
77271-848
77747-912
6-Chloropurine 98%
6-Chloropurine
Caution: for Research Use Only
Formula:
C₅H₃ClN₄ MW: 154.56 g/mol |
MDL Number:
MFCD00075825 CAS Number: 87-42-3 Merck Index: 13,02180 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...