[Oxalylbis(azanediyl)]bis(ethane-2,1-diyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate] 97%

Supplier: Ambeed
Danger

Synonyms: Antioxidant MD-697, (Oxalylbis(azanediyl))bis(ethane-2,1-diyl) bis(3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate)

A122696-100G A122696-10G A122696-1G A122696-5G A122696-500G
77271-596EA 216 USD
77271-596 77271-598 77271-600 77271-602 77271-604 77566-330
[Oxalylbis(azanediyl)]bis(ethane-2,1-diyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate] 97%
[Oxalylbis(azanediyl)]bis(ethane-2,1-diyl) bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate]

Caution: For research use only.

Formula: C₄₀H₆₀N₂O₈
MW: 696.93 g/mol
MDL Number: MFCD01940880
CAS Number: 70331-94-1
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR