3,4-Diaminotoluene 97%

Supplier: Ambeed
Warning

A119641-100G A119641-25G A119641-500G
77271-418EA 15 USD
77271-418 77271-420 77271-422
3,4-Diaminotoluene 97%
3,4-Diaminotoluene

Caution: for Research Use Only

Formula: C₇H₁₀N₂
MDL Number: MFCD00007728
CAS Number: 496-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR