Z-D-Lys(Boc)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Cbz-D-Lys(Boc)-OH, Nα-Z-Nε-Boc-D-lysine, Nα-Cbz-Nε-Boc-D-Lysine
A112843-100G
A112843-10G
A112843-1G
A112843-250MG
A112843-5G
A112843-25G
77270-994EA
336
USD
77270-994
77270-996
77270-998
77271-000
77271-002
77570-706
Z-D-Lys(Boc)-OH 97%
Z-D-Lys(Boc)-OH
Caution: for Research Use Only
Formula:
C₁₉H₂₈N₂O₆ MW: 380.441 g/mol |
MDL Number:
MFCD00065699 CAS Number: 66845-42-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...