2,2'-O-Cyclocytidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride

A112401-100G A112401-1G A112401-1MG A112401-250MG A112401-25G A112401-5G
77245-758EA 234 USD
77245-758 77245-760 77245-762 77245-764 77245-766 77245-768
2,2'-O-Cyclocytidine hydrochloride 97%
2,2'-O-Cyclocytidine hydrochloride

Caution: for Research Use Only

Formula: C₉H₁₁N₃O₄
MW: 261.66 g/mol
MDL Number: MFCD00012636
CAS Number: 10212-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR