2-Bromo-4,5-difluorophenol 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-3,4-difluoro-6-hydroxybenzene

A109988-1G A109988-5G A109988-25G
77283-916EA 9 USD
77283-916 77283-918 77570-560
2-Bromo-4,5-difluorophenol 98%
2-Bromo-4,5-difluorophenol

Caution: for Research Use Only

Formula: C₆H₃BrF₂O
MDL Number: MFCD00070751
CAS Number: 166281-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR