3-Bromo-4-ethoxybenzoic acid 95%

Supplier: Ambeed
Warning

A108133-100MG A108133-1G A108133-250MG
77270-688EA 12 USD
77270-688 77270-690 77270-692
3-Bromo-4-ethoxybenzoic acid 95%
3-Bromo-4-ethoxybenzoic acid

Caution: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.08 g/mol
MDL Number: MFCD01143606
CAS Number: 24507-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR