2-Methyl-2-adamantanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-2-methyladamantane, 2-Methyladamantan-2-ol

A107840-1G A107840-25G A107840-5G A107840-100G
77257-504EA 9 USD
77257-504 77257-506 77257-508 77549-078
2-Methyl-2-adamantanol 97%
2-Methyl-2-adamantanol

Caution: for Research Use Only

Formula: C₁₁H₁₈O
MW: 166.26 g/mol
MDL Number: MFCD00074811
CAS Number: 702-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR