4-(Boc-amino)-2-chloropyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-(tert-butoxycarbonyl)pyridine, tert-Butyl (2-chloropyridin-4-yl)carbamate, 4-Amino-2-chloropyridine, 4-BOC protected, tert-Butyl 2-chloropyridine-4-carbamate

A107480-1G A107480-250MG A107480-5G
77283-610EA 5.15 USD
77283-610 77283-612 77283-614 77283-616
4-(Boc-amino)-2-chloropyridine 95%
4-(Boc-amino)-2-chloropyridine

Caution: for Research Use Only

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD02179232
CAS Number: 234108-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR