N-Methyl-2-nitroaniline 98%

Supplier: Ambeed
Danger

A985375-10G A985375-25G A985375-5G
77200-484EA 19 USD
77200-484 77200-486 77200-488
N-Methyl-2-nitroaniline 98%
N-Methyl-2-nitroaniline

Caution: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD00007090
CAS Number: 612-28-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR