1,6-Hexanedithiol 98%

Supplier: Ambeed
Danger

Synonyms: 1,6-Dimercaptohexane

A808980-5G A808980-25G
77199-678EA 17 USD
77199-678 77573-584
1,6-Hexanedithiol 98%
1,6-Hexanedithiol

Caution: for Research Use Only

Formula: C₆H₁₄S₂
MW: 150.31 g/mol
Boiling Pt: 242
Melting Pt: -21
MDL Number: MFCD00004910
CAS Number: 1191-43-1
UN: 3334
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR