1,2-Diiodotetrafluorobenzene 97%

Supplier: Ambeed
Warning

A661546-1G A661546-250MG A661546-25G A661546-5G
77198-516EA 20.6 USD
77198-516 77198-518 77198-520 77198-522
1,2-Diiodotetrafluorobenzene 97%
1,2-Diiodotetrafluorobenzene

Caution: for Research Use Only

Formula: C₆F₄I₂
MW: 401.87 g/mol
MDL Number: MFCD00019011
CAS Number: 2708-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR