(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A558866-100MG
A558866-1G
A558866-250MG
A558866-5G
77197-580EA
14
USD
77197-580
77197-582
77197-584
77197-586
(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) ≥98%, ee 99%
(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline)
Caution: for Research Use Only
Formula:
C₂₁H₂₂N₂O₂ |
MDL Number:
MFCD00192245 CAS Number: 131457-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...