4-Methoxycyclohexene-1-boronic Acid Pinacol Ester 98%

Supplier: Ambeed
Warning

A505686-250MG A505686-1G A505686-5G
77169-392EA 92 USD
77169-392 77893-017 77893-018
4-Methoxycyclohexene-1-boronic Acid Pinacol Ester 98%
4-Methoxycyclohexene-1-boronic Acid Pinacol Ester

Caution: for Research Use Only

Formula: C₁₃H₂₃BO₃
MDL Number: MFCD18383325
CAS Number: 1092938-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR