1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose 95%

Supplier: Ambeed
Warning

Synonyms: Diacetone-D-galactose, 1,2:3,4-Di-O-isopropylidene-D-galactopyranose, ((3aR,5R,5aS,8aS,8bR)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-5-yl)methanol

A466452-100G A466452-1G A466452-25G A466452-5G
77196-634EA 436.72 USD
77196-634 77196-636 77196-638 77196-640
1,2:3,4-Di-O-isopropylidene-ɑ-D-galactopyranose 95%
1,2:3,4-Di-O-isopropylidene-alpha-D-galactopyranose

Caution: for Research Use Only

Formula: C₁₂H₂₀O₆
MW: 260.29 g/mol
MDL Number: MFCD00063225
CAS Number: 4064-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR