trans-1,4-Cyclohexanedicarboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A206067-10G
A206067-25G
A206067-5G
A206067-100G
77192-488EA
6
USD
77192-488
77192-490
77192-492
77593-344
trans-1,4-Cyclohexanedicarboxylic acid 97%
trans-1,4-Cyclohexanedicarboxylic acid
Caution: for Research Use Only
Formula:
C₈H₁₂O₄ |
MDL Number:
MFCD00066203 CAS Number: 619-82-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...