3-Bromothiophenol 98%, Ambeed.Inc

Supplier: Ambeed
Danger

Synonyms: m-Bromothiophenol, 3-Bromobenzenethiol, 1-Bromo-3-mercaptobenzene, 3-Bromobenzene-1-thiol

A199047-1G A199047-5G A199047-25G A199047-100G
77164-526EA 8.24 USD
77164-526 77164-528 77550-264 77683-551
3-Bromothiophenol 98%, Ambeed.Inc
3-Bromothiophenol

Caution: For research use only.

Formula: C₆H₅BrS
MW: 189.08 g/mol
MDL Number: MFCD00009603
CAS Number: 6320-01-0
UN: 3334
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR