2-Amino-4-(trifluoromethyl)oxazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Trifluoromethyl)oxazol-2-amine
A163237-1G
A163237-250MG
A163237-5G
A163237-100MG
77163-256EA
44
USD
77163-256
77163-258
77163-260
77892-104
2-Amino-4-(trifluoromethyl)oxazole 97%
2-Amino-4-(trifluoromethyl)oxazole
Caution: for Research Use Only
Formula:
C₄H₃F₃N₂O |
MDL Number:
MFCD07777221 CAS Number: 35629-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...